Learn More
Biomatik Corporation Allopregnanolone (AP) ELISA Kit
Supplier:  Biomatik Corporation EKU02257
Encompass_Preferred
Allopregnanolone (AP) ELISA Kit for detecting AP in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.51ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: THP; 3a,5a-Tetrahydro Progesterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5779