Learn More
Biomatik Corporation Human Tryptophanyl tRNA Synthetase (WARS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08602
Encompass_Preferred
Human Tryptophanyl tRNA Synthetase (WARS) ELISA Kit for detecting WARS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23381. Gene ID: 7453. Target Synonyms: GAMMA-2; IFI53; IFP53; WRS; TrpRS; hWRS; Tryptophan tRNA Ligase 1,Cytoplasmic; Interferon-induced protein 53; Tryptophanyl-tRNA synthetase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4252