Learn More
Biomatik Corporation Horse Alpha-Fetoprotein (AFP) ELISA Kit
Supplier:  Biomatik Corporation EKU02290
Encompass_Preferred
Horse Alpha-Fetoprotein (AFP) ELISA Kit for detecting AFP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 6.25-400pg/ml. Sensitivity: 2.59pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49066. Gene ID: 100034185. Target Synonyms: aFP; A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5781