Learn More
Biomatik Corporation Human Activin A (ACVA) ELISA Kit, 96 tests
Catalog No. 501504254
Encompass_Preferred
The Human Activin A (ACVA) ELISA Kit allows for quantitative determination of Activin A in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.63 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P08476. Cross Activity: No significant cross-reactivity or interference between Activin A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4254 Supplier Biomatik Corporation Supplier No. EKU08665
The Human Activin A (ACVA) ELISA Kit allows for quantitative determination of Activin A in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.63 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P08476. Cross Activity: No significant cross-reactivity or interference between Activin A and analogues was observed. Restriction: For Research Use Only.