Learn More
Biomatik Corporation Cyclic Adenosine Monophosphate (cAMP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08667
Encompass_Preferred
The Cyclic Adenosine Monophosphate (cAMP) ELISA Kit allows for quantitative determination of Cyclic Adenosine Monophosphate in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 246.9-20000 pg/mL. Sensitivity: 92.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Cyclic Adenosine Monophosphate and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4255