Learn More
Biomatik Corporation Pig Insulin (INS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08683
Encompass_Preferred
Pig Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 61.7-5,000pg/ml. Sensitivity: 23.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01315. Gene ID: 397415. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4258