Learn More
Biomatik Corporation Human Antithrombin (AT) ELISA Kit
Supplier:  Biomatik Corporation EKU02467
Encompass_Preferred
Human Antithrombin (AT) ELISA Kit for detecting AT in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01008. Gene ID: 462. Target Synonyms: AT3; ATIII; SERPINC1; Anti-Thrombin Antibodies; Serpin Peptidase Inhibitor Clade C Member 1; Coding Sequence Signal Peptide Antithrombin Part 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5807