Learn More
Biomatik Corporation Cattle Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit, 96 tests
Catalog No. 501504261
Encompass_Preferred
The Cattle Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 2, Basic in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.49 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P03969. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 2, Basic and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4261 Supplier Biomatik Corporation Supplier No. EKU08689
The Cattle Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 2, Basic in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.49 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P03969. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 2, Basic and analogues was observed. Restriction: For Research Use Only.