Learn More
Biomatik Corporation Human 11-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD11b1) ELISA Kit
Supplier:  Biomatik Corporation EKU02002
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.56ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P28845. Target Synonyms: 11DH; 11-beta-HSD1; HDL; HSD11; HSD11B; HSD11L; SDR26C1; Corticosteroid 11-beta-dehydrogenase isozyme 1; Short Chain Dehydrogenase/Reductase Family 26C,Member 1. Restriction: For Research Use Only.
Catalog No. 50-228-5747