Learn More
Biomatik Corporation 5-Hydroxyindoleacetic Acid (5-HIAA) ELISA Kit
Supplier:  Biomatik Corporation EKU02026
Encompass_Preferred
Sample Type: serum, plasma, urine, cerebrospinal fluid and other biological fluids. Species Reactivity: General species. Detection Range: 4.94-400ng/mL. Sensitivity: 1.93ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Competitive. Assay Time: 2 hours. UniProt ID: Not Available. Target Synonyms: 5HIAA. Restriction: For Research Use Only.
Catalog No. 50-228-5753