Learn More
Biomatik Corporation 8-Epi Prostaglandin F2 Alpha (8-EpiPGF2-α) ELISA Kit
Supplier:  Biomatik Corporation EKU02038
Encompass_Preferred
8-Epi Prostaglandin F2 Alpha (8-EpiPGF2-α) ELISA Kit for detecting 8-EpiPGF2-α in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.73pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 8-EpiPGF2-α; 8epi-PGF2a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5755