Learn More
Biomatik Corporation Human Acid Sphingomyelinase (ASM) ELISA Kit
Supplier:  Biomatik Corporation EKU02086
Encompass_Preferred
Human Acid Sphingomyelinase (ASM) ELISA Kit for detecting ASM in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17405. Gene ID: 6609. Target Synonyms: SMPD1; NPD; SMase; aSMase; Sphingomyelin Phosphodiesterase 1,Acid Lysosomal; Simply Sphingomyelinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5759