Learn More
Biomatik Corporation Mouse Von Willebrand Factor (vWF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08707
Encompass_Preferred
Mouse Von Willebrand Factor (vWF) ELISA Kit for detecting vWF in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 4.82ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8CIZ8. Gene ID: 22371. Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4266