Learn More
Biomatik Corporation Pig Adrenomedullin (ADM) ELISA Kit
Supplier:  Biomatik Corporation EKU02187
Encompass_Preferred
Pig Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.21pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P53366. Gene ID: 397195. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5768