Learn More
Biomatik Corporation Human Aldolase A, Fructose Bisphosphate (ALDOA) ELISA Kit
Supplier:  Biomatik Corporation EKU02230
Encompass_Preferred
Human Aldolase A, Fructose Bisphosphate (ALDOA) ELISA Kit for detecting ALDOA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04075. Gene ID: 226. Target Synonyms: FBA; ALDO-A; Fructose 1,6 Bisphosphate Aldolase; Lung cancer antigen NY-LU-1; Muscle-type aldolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5773