Learn More
Biomatik Corporation Human Alkaline Phosphatase, Placental Like Protein 2 (ALPPL2) ELISA Kit
Supplier:  Biomatik Corporation EKU02250
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P10696. Target Synonyms: ALPG; GCAP; ALP-1; Alkaline phosphatase Nagao isozyme; Germ cell alkaline phosphatase; Placental alkaline phosphatase-like. Restriction: For Research Use Only.
Catalog No. 50-228-5778