Learn More
Biomatik Corporation Human Alpha 2-Antiplasmin (a2PI) ELISA Kit
Supplier:  Biomatik Corporation EKU02282
Encompass_Preferred
Human Alpha 2-Antiplasmin (a2PI) ELISA Kit for detecting a2PI in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 4.7-300ng/ml. Sensitivity: 1.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08697. Gene ID: 5345. Target Synonyms: SERPINF2; AAP; A2-PI; API; PLI; A2AP; Alpha-2-Plasmin Inhibitor; Serpin Peptidase Inhibitor Clade F Member 2; Pigment Epithelium Derived Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5780