Learn More
Biomatik Corporation Rabbit Serum Amyloid A (SAA) ELISA Kit, 96 tests
Catalog No. 501504271
Encompass_Preferred
Rabbit Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 25-400ng/ml. Sensitivity: 10ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P53614. Gene ID: 100009168. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4271 Supplier Biomatik Corporation Supplier No. EKU08714
Rabbit Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 25-400ng/ml. Sensitivity: 10ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P53614. Gene ID: 100009168. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.