Learn More
Biomatik Corporation Mouse Myostatin (MSTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08728
Encompass_Preferred
Mouse Myostatin (MSTN) ELISA Kit for detecting MSTN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 617.3-50000pg/ml. Sensitivity: 253.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O08689. Gene ID: 17700. Target Synonyms: GDF8; Growth Differentiation Factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4274