Learn More
Biomatik Corporation Rat Hepcidin (Hepc) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08734
Encompass_Preferred
Rat Hepcidin (Hepc) ELISA Kit for detecting Hepc in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 160-100,000pg/ml. Sensitivity: 57pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q99MH3. Gene ID: 84604. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4276