Learn More
Biomatik Corporation Rat Antidiuretic Hormone (ADH) ELISA Kit
Supplier:  Biomatik Corporation EKU02448
Encompass_Preferred
Rat Antidiuretic Hormone (ADH) ELISA Kit for detecting ADH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.43pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01186. Gene ID: 24221. Target Synonyms: ARVP; AVP; VP; Arginine Vasopressin; Arg-Vasopressin; Antidiuretic Hormone; Diabetes Insipidus; Neurohypophyseal; Argipressin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5804