Learn More
Biomatik Corporation Human Ras Related C3 Botulinum Toxin Substrate 1 (Rac1) ELISA Kit
Supplier:  Biomatik Corporation EKU07013
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P63000. Target Synonyms: TC-25; p21-Rac1; MIG5; Cell migration-inducing gene 5 protein; Ras-like protein TC25; p21-Rac1. Restriction: For Research Use Only.
Catalog No. 50-228-6350