Learn More
Biomatik Corporation Human Receptor Activator Of Nuclear Factor Kappa B (RANk) ELISA Kit
Supplier:  Biomatik Corporation EKU07017
Encompass_Preferred
Human Receptor Activator Of Nuclear Factor Kappa B (RANk) ELISA Kit for detecting RANk in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y6Q6. Gene ID: 8792. Target Synonyms: CD265; TNFRSF11A; TNFRSF11-A; ODFR; OFE; TRANCER; Osteoclast differentiation factor receptor; Tumor Necrosis Factor Receptor Superfamily,Member 11a,NFKB Activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6352