Learn More
Biomatik Corporation Kynurenic Acid (KYNA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08748
Encompass_Preferred
Kynurenic Acid (KYNA) ELISA Kit for detecting KYNA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ng/ml. Sensitivity: 0.89ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Kinurenic acid; Kynuronic acid; Kuinurenic acid; Transtorine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4282