Learn More
Biomatik Corporation Salmon Calcitonin (SCT) ELISA Kit
Supplier:  Biomatik Corporation EKU07189
Encompass_Preferred
Salmon Calcitonin (SCT) ELISA Kit for detecting SCT in biological agents. Species Reactivity: Universal. Detection Range: 78.1-20,000pg/ml. Sensitivity: 31.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: CGRP; Salcatonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6366