Learn More
Biomatik Corporation Homocysteine (HCy) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08750
Encompass_Preferred
The Homocysteine (HCy) ELISA Kit allows for quantitative determination of Homocysteine in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 98.77-8000 ng/mL. Sensitivity: 39.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Homocysteine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4283