Learn More
Biomatik Corporation Quinolinic Acid (QA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08752
Encompass_Preferred
Quinolinic Acid (QA) ELISA Kit for detecting QA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: QUIN; Pyridine-2,3-Dicarboxylic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4284