Learn More
Biomatik Corporation Docosahexaenoic Acid (DHA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08761
Encompass_Preferred
The Docosahexaenoic Acid (DHA) ELISA Kit allows for quantitative determination of Docosahexaenoic Acid in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.65 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Docosahexaenoic Acid and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4288