Learn More
Biomatik Corporation Human TANK Binding Kinase 1 (TBK1) ELISA Kit
Supplier:  Biomatik Corporation EKU07568
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9UHD2. Target Synonyms: NAK; T2K; NF-kappa-B-activating kinase; Serine/threonine-protein kinase TBK1. Restriction: For Research Use Only.
Catalog No. 50-228-6402