Learn More
Biomatik Corporation Mouse Thioredoxin-interacting Protein (TXNIP) ELISA Kit
Supplier:  Biomatik Corporation EKU07618
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8BG60. Target Synonyms: TBP2; HHCPA78; THIF; VDUP1;Thioredoxin Binding Protein 2 ; Vitamin D3-Upregulated Protein 1; Upregulated By 1,25-Dihydroxyvitamin D-3. Restriction: For Research Use Only.
Catalog No. 50-228-6405