Learn More
Biomatik Corporation Human Thyroid Transcription Factor 1 (TITF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07686
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.058ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P43699. Target Synonyms: TTF1; NKX2-1; BCH; BHC; NK-2; NKX2.1; NKX2A; TEBP; NK2 Homeobox 1; Benign Chorea; Thyroid nuclear factor 1; Thyroid-specific enhancer-binding protein. Restriction: For Research Use Only.
Catalog No. 50-228-6417