Learn More
Biomatik Corporation Human Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU08818
Encompass_Preferred
The Human Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit (CLIA) allows for quantitative determination of Monocyte Chemotactic Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.74-2000 pg/mL. Sensitivity: 0.97 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P13500. Cross Activity: No significant cross-reactivity or interference between Monocyte Chemotactic Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4296