Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha (TNFa) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU08843
Encompass_Preferred
Mouse Tumor Necrosis Factor Alpha (TNFa) CLIA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06804. Gene ID: 21926. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4298