Learn More
Biomatik Corporation Human Procalcitonin (PCT) ELISA Kit (CLIA), 96 tests
Catalog No. 501504301
Encompass_Preferred
The Human Procalcitonin (PCT) ELISA Kit (CLIA) allows for quantitative determination of Procalcitonin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.86-5,000 pg/mL. Sensitivity: 2.75 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P01258. Cross Activity: No significant cross-reactivity or interference between Procalcitonin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4301 Supplier Biomatik Corporation Supplier No. EKU08908
The Human Procalcitonin (PCT) ELISA Kit (CLIA) allows for quantitative determination of Procalcitonin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.86-5,000 pg/mL. Sensitivity: 2.75 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P01258. Cross Activity: No significant cross-reactivity or interference between Procalcitonin and analogues was observed. Restriction: For Research Use Only.