Learn More
Biomatik Corporation Gallus Lysozyme (LZM) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU08933
Encompass_Preferred
The Gallus Lysozyme (LZM) ELISA Kit (CLIA) allows for quantitative determination of Lysozyme in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 13.7-10000 pg/mL. Sensitivity: 5.4 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P00698. Cross Activity: No significant cross-reactivity or interference between Lysozyme and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4302