Learn More
Biomatik Corporation Human Thymic Stromal Lymphopoietin (TSLP) ELISA Kit (CLIA), 96 tests
Catalog No. 501504303
Encompass_Preferred
Human Thymic Stromal Lymphopoietin (TSLP) CLIA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.13pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q969D9. Gene ID: 85480. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4303 Supplier Biomatik Corporation Supplier No. EKU08937
Human Thymic Stromal Lymphopoietin (TSLP) CLIA Kit for detecting TSLP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.13pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q969D9. Gene ID: 85480. Restrictions: For Research Use Only. Not for use in diagnostic procedures.