Learn More
Biomatik Corporation Gallus Interleukin 18 (IL18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08970
Encompass_Preferred
The Gallus Interleukin 18 (IL18) ELISA Kit allows for quantitative determination interleukin 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8QFQ8. Cross Activity: No significant cross-reactivity or interference between Interleukin 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4308