Learn More
Biomatik Corporation Multi-species Ubiquitin (Ub) ELISA Kit, 96 tests
Catalog No. 501504311
Encompass_Preferred
Human/Mouse/Rat/Rabbit/Dog/Pig/Bovine/Horse Ubiquitin (Ub) ELISA Kit for detecting Ub in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Horse (Equus caballus; Equine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.42ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0CG47. Gene ID: 7314. Target Synonyms: Ubiquitin B; UbB; Ubiquitous Immunopoietic Polypeptide; Polyubiquitin B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4311 Supplier Biomatik Corporation Supplier No. EKU08985
Human/Mouse/Rat/Rabbit/Dog/Pig/Bovine/Horse Ubiquitin (Ub) ELISA Kit for detecting Ub in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Horse (Equus caballus; Equine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.42ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0CG47. Gene ID: 7314. Target Synonyms: Ubiquitin B; UbB; Ubiquitous Immunopoietic Polypeptide; Polyubiquitin B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.