Learn More
Biomatik Corporation Mouse Caveolin 1 (CAV1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08988
Encompass_Preferred
Mouse Caveolin 1 (CAV1) ELISA Kit for detecting CAV1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49817. Gene ID: 12389. Target Synonyms: VIP21; Caveolae Protein 22kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4312