Learn More
Biomatik Corporation Mouse Alpha-1-Microglobulin/Bikunin Precursor (a1M) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08989
Encompass_Preferred
Mouse Alpha-1-Microglobulin (a1M) ELISA Kit for detecting a1M in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07456. Gene ID: 11699. Target Synonyms: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4313