Learn More
Biomatik Corporation Dog Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKU08383
Encompass_Preferred
Dog Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 2.47-200ng/ml. Sensitivity: 0.91ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9XSW8; E2RJZ8. Gene ID: 403483; 485428. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6495