Learn More
Biomatik Corporation Rat Mucin 5 Subtype B (MUC5B) ELISA Kit, 96 tests
Catalog No. 501504319
Encompass_Preferred
Rat Mucin 5 Subtype B (MUC5B) ELISA Kit for detecting MUC5B in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZAU0. Target Synonyms: MUC5-B; MUC9; MG1; Oligomeric Mucus/Gel-Forming; Mucin 5,Subtype B,Tracheobronchial; Cervical mucin; High molecular weight salivary mucin MG1; Sublingual gland mucin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4319 Supplier Biomatik Corporation Supplier No. EKU09017
Rat Mucin 5 Subtype B (MUC5B) ELISA Kit for detecting MUC5B in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZAU0. Target Synonyms: MUC5-B; MUC9; MG1; Oligomeric Mucus/Gel-Forming; Mucin 5,Subtype B,Tracheobronchial; Cervical mucin; High molecular weight salivary mucin MG1; Sublingual gland mucin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.