Learn More
Biomatik Corporation Mouse Complement 1q (C1q) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09021
Encompass_Preferred
Mouse Complement 1q (C1q) ELISA Kit for detecting C1q in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.244ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98086; P14106; Q02105. Gene ID: 12259; 12260; 12262. Target Synonyms: C1-q; Autoantigen Clq. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4320