Learn More
Biomatik Corporation Rat Complement 1q (C1q) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09022
Encompass_Preferred
Rat Complement 1q (C1q) ELISA Kit for detecting C1q in serum, plasma,, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31720; P31721; P31722. Gene ID: 298566; 29687; 362634. Target Synonyms: C1-q; Autoantigen Clq. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4321