Learn More
Biomatik Corporation Rat Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09025
Encompass_Preferred
Rat Alpha-1-Acid Glycoprotein (a1AGP) ELISA Kit for detecting a1AGP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000ng/ml. Sensitivity: 14.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02764. Gene ID: 24614. Target Synonyms: AGP1; a1-AGP; ORM1; OMD 1; Orosomucoid 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4322