Learn More
Biomatik Corporation Mouse Complement Component 4 (C4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09034
Encompass_Preferred
Mouse Complement Component 4 (C4) ELISA Kit for detecting C4 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500ng/ml. Sensitivity: 2.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01029. Gene ID: 12268. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4325