Learn More
Biomatik Corporation Mouse Chemokine C-C-Motif Receptor 2 (CCR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09035
Encompass_Preferred
Mouse Chemokine C-C-Motif Receptor 2 (CCR2) ELISA Kit for detecting CCR2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51683. Gene ID: 12772. Target Synonyms: CD192; CC-CKR2; CCR2A; C-C CKR-2; CCR2B; CKR2-A; CKR2-B; CMKBR2; MCP1-R; Monocyte chemoattractant protein 1 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4326