Learn More
Biomatik Corporation Phosphatidylserine (PS) ELISA Kit
Supplier:  Biomatik Corporation EKU08732
Encompass_Preferred
Phosphatidylserine (PS) ELISA Kit for detecting PS in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 14.81-1,200ng/ml. Sensitivity: 5.03ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Ptd-L-Ser; PtdSer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6536