Learn More
Biomatik Corporation Human Kallikrein 7 (KLK7) CLIA Kit
Supplier:  Biomatik Corporation EKU08947
Encompass_Preferred
Human Kallikrein 7 (KLK7) CLIA Kit for detecting KLK7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P49862. Gene ID: 5650. Target Synonyms: PRSS6; SCCE; hK7; Kallikrein-Related Peptidase 7; Serine protease 6; Stratum corneum chymotryptic enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6555