Learn More
Biomatik Corporation Rat Cytochrome P450 3A1 (CYP3A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09050
Encompass_Preferred
Rat Cytochrome P450 3A1 (CYP3A1) ELISA Kit for detecting CYP3A1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04800. Target Synonyms: CYPIIIA1; Cytochrome P450-PCN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4332